3-Hydroxy-4-nitrobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

156167-10G 156167-1G
SIAL156167-10GEA 274 EUR
SIAL156167-10G SIAL156167-1G
3-Hydroxy-4-nitrobenzaldehyde, Sigma-Aldrich®
3-Hydroxy-4-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007109
CAS Number: 704-13-2
EINECS: 211-879-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR