4-Hydroxy-3-nitrobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
144320-25G 144320-5G
SIAL144320-25GEA 56.4 EUR
SIAL144320-25G SIAL144320-5G
4-Hydroxy-3-nitrobenzaldehyde, Sigma-Aldrich®
4-Hydroxy-3-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007117
CAS Number: 3011-34-5
EINECS: 221-141-6