o-Anisaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde

MQ300

W407701-10KG-K W407701-1KG-K W407701-5KG-K
SIALW407701-10KG-KEA 1010 EUR
SIALW407701-10KG-K SIALW407701-1KG-K SIALW407701-5KG-K
o-Anisaldehyde, Sigma-Aldrich®
o-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 238
Melting Pt: 34…37 °C
Density: 1.127 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4
EINECS: 205-171-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR