m-Anisaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methoxybenzaldehyde
M-Clarity™ quality level = MQ200
129658-100G 129658-25G
SIAL129658-100GEA 63 EUR
SIAL129658-100G SIAL129658-25G
m-Anisaldehyde, Sigma-Aldrich®
m-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 228…229 °C (1013 hPa)
Density: 1.1149 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8