4-Ethoxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
173606-100G
173606-25G
SIAL173606-100GEA
107
EUR
SIAL173606-100G
SIAL173606-25G
4-Ethoxybenzaldehyde, Sigma-Aldrich®
4-Ethoxybenzaldehyde
Formula:
C₉H₁₀O₂ Boiling Pt: 255 °C (1013 hPa) Melting Pt: 13…14 °C Density: 1.0509 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003388 CAS Number: 10031-82-0 EINECS: 233-093-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...