p-Anisaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methoxybenzaldehyde
M-Clarity™ quality level = MQ200
A88107-100G A88107-1KG A88107-500G A88107-5G
SIALA88107-100GEA 46 EUR
SIALA88107-100G SIALA88107-1KG SIALA88107-500G SIALA88107-5G
p-Anisaldehyde, Sigma-Aldrich®
p-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140
Melting Pt: 0…2 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666