p-Anisaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methoxybenzaldehyde
W267007-1KG-K W267007-25KG-K W267007-5KG-K
SIALW267007-1KG-KEA 90 EUR
SIALW267007-1KG-K SIALW267007-25KG-K SIALW267007-5KG-K
p-Anisaldehyde, Sigma-Aldrich®
p-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140
Melting Pt: 0…2 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666