2-Hydroxy-m-anisaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms:
o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde
M-Clarity™ quality level = MQ100
120804-10G
120804-500G
SIAL120804-10GEA
41.1
EUR
SIAL120804-10G
SIAL120804-500G
2-Hydroxy-m-anisaldehyde, Sigma-Aldrich®
2-Hydroxy-m-anisaldehyde
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 265 Melting Pt: 40…42 °C Density: 1.2039 g/cm³ (45 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003322 CAS Number: 148-53-8 EINECS: 205-715-3 |