Vanillin, Sigma-Aldrich®
Supplier: Merck
Warning
V1104-100G V1104-2G V1104-500G V1104-2KG
SIALV1104-100GEA 43.8 EUR
SIALV1104-100G SIALV1104-2G SIALV1104-500G SIALV1104-2KG
Vanillin, Sigma-Aldrich®
Vanillin
Formula: C₈H₈O₃
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: ∼82 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM