Veratraldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,4-Dimethoxybenzaldehyde
MQ100
143758-100G 143758-500G 143758-5G
SIAL143758-100GEA 59.2 EUR
SIAL143758-100G SIAL143758-500G SIAL143758-5G
Veratraldehyde, Sigma-Aldrich®
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281
Melting Pt: 40…43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084