Veratraldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,4-Dimethoxybenzaldehyde
M-Clarity™ quality level = MQ100
W310905-10KG-K W310905-1KG-K W310905-25KG-K W310905-5KG-K
SIALW310905-10KG-KEA 963 EUR
SIALW310905-10KG-K SIALW310905-1KG-K SIALW310905-25KG-K SIALW310905-5KG-K
Veratraldehyde, Sigma-Aldrich®
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 40…43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084