3-Hydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
H19808-100G H19808-500G H19808-5G
SIALH19808-100GEA 58.8 EUR
SIALH19808-100G SIALH19808-500G SIALH19808-5G
3-Hydroxybenzaldehyde, Sigma-Aldrich®
3-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 241
Melting Pt: 99…102 °C
Density: 1.1299 g/cm³ (114 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9