4-Hydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
MQ300
W398403-10KG-K
W398403-1KG-K
W398403-5KG-K
SIALW398403-10KG-KEA
1150
EUR
SIALW398403-10KG-K
SIALW398403-1KG-K
SIALW398403-5KG-K
4-Hydroxybenzaldehyde, Sigma-Aldrich®
4-Hydroxybenzaldehyde
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 310 Melting Pt: 116 Density: 1.13 g/cm³ (20 °C) Flash Pt: 174 Storage Temperature: Ambient |
MDL Number:
MFCD00006939 CAS Number: 123-08-0 EINECS: 204-599-1 Merck Index: 13,04836 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...