2,3-Dihydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
189839-25G 189839-5G
SIAL189839-25GEA 173 EUR
SIAL189839-25G SIAL189839-5G
2,3-Dihydroxybenzaldehyde, Sigma-Aldrich®
2,3-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1