2,4-Dihydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
168637-100G 168637-25G
SIAL168637-100GEA 92.5 EUR
SIAL168637-100G SIAL168637-25G
2,4-Dihydroxybenzaldehyde, Sigma-Aldrich®
2,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 351
Melting Pt: 132…133 °C
Density: 1.448 g/cm³ (–93 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 12,08324