3,4-Dihydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Protocatechuic aldehyde, Protocatechualdehyde
M-Clarity™ quality level = MQ100
D108405-100G D108405-25G D108405-5G
SIALD108405-100GEA 131 EUR
SIALD108405-100G SIALD108405-25G SIALD108405-5G
3,4-Dihydroxybenzaldehyde, Sigma-Aldrich®
3,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 150…155 °C
Density: 1.505 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 12,08079