4-(Methylthio)benzaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
222771-10G 222771-50G
SIAL222771-10GEA 42 EUR
SIAL222771-10G SIAL222771-50G
4-(Methylthio)benzaldehyde, Sigma-Aldrich®
4-(Methylthio)benzaldehyde
Formula: C₈H₈OS
MW: 152.22 g/mol
Boiling Pt: 267 °C (1013 hPa)
Density: 1.14 g/cm³ (20 °C)
MDL Number: MFCD00006948
CAS Number: 3446-89-7
EINECS: 222-365-7
UN: 3334
ADR: 9,