Terephthalaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

T2207-100G T2207-500G T2207-5G
SIALT2207-100GEA 66.4 EUR
SIALT2207-100G SIALT2207-500G SIALT2207-5G
Terephthalaldehyde, Sigma-Aldrich®
Terephthalaldehyde
Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Density: 1.381 g/cm³ (20 °C)
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR