1,2,3-Trichlorobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
T54402-100G T54402-500G T54402-5G
SIALT54402-100GEA 47.2 EUR
SIALT54402-100G SIALT54402-500G SIALT54402-5G
1,2,3-Trichlorobenzene, Sigma-Aldrich®
1,2,3-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 220…222 °C (1013 hPa)
Melting Pt: 53…54 °C
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000537
CAS Number: 87-61-6
EINECS: 201-757-1
UN: 3077
ADR: 9,III
Merck Index: 12,09759