1,2,4-Trichlorobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
296104-100ML 296104-1L
SIAL296104-100MLEA 93.1 EUR
SIAL296104-100ML SIAL296104-1L
1,2,4-Trichlorobenzene, Sigma-Aldrich®
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4
Melting Pt: 17
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1 III
Merck Index: 13,09704