Chlorobenzene HPLC
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene
H27688.AU H27688.0F
H27688.AUEA 139 EUR
H27688.AU H27688.0F
Chlorobenzene HPLC
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 130
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3 III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM