Anti- Goat Polyclonal Antibody (Unconjugated)

Supplier: Immunoreagents
IMMRIR2371EA 52.6 EUR
IMMRIR2371
Anti- Goat Polyclonal Antibody (Unconjugated)
Antibodies
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR