Anti-IgG Dog Polyclonal Antibody (Unconjugated)

Supplier: Immunoreagents
IMMRIR2379EA 133 EUR
IMMRIR2379
Anti-IgG Dog Polyclonal Antibody (Unconjugated)
Antibodies
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Dog
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR