3-Chlorophenylboronic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane
417521-10G
417521-1G
SIAL417521-10GEA
169
EUR
SIAL417521-10G
SIAL417521-1G
3-Chlorophenylboronic acid, Sigma-Aldrich®
3-Chlorophenylboronic acid
Formula:
C₆H₆BClO₂ MW: 156.38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00161354 CAS Number: 63503-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...