2,4-Dichlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
146757-100G 146757-5G
SIAL146757-100GEA 55.4 EUR
SIAL146757-100G SIAL146757-5G
2,4-Dichlorobenzaldehyde, Sigma-Aldrich®
2,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 68…71 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS Number: 874-42-0
EINECS: 212-861-1
UN: 3077
ADR: 9,III