3,4-Dichlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
D56608-25G D56608-5G
SIALD56608-25GEA 68.6 EUR
SIALD56608-25G SIALD56608-5G
3,4-Dichlorobenzaldehyde, Sigma-Aldrich®
3,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003351
CAS Number: 6287-38-3
EINECS: 228-520-5
UN: 1759
ADR: 8,II