4-Chlorobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: p-Chlorobenzonitrile

M-Clarity™ quality level = MQ100

115622-100G 115622-25G
SIAL115622-100GEA 265 EUR
SIAL115622-100G SIAL115622-25G
4-Chlorobenzonitrile, Sigma-Aldrich®
4-Chlorobenzonitrile
Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 223
Melting Pt: 90…93 °C
Density: 1.1133 g/cm³ (16.8 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR