2,4,6-Trichlorobenzonitrile

Supplier: Thermo Scientific

TL00272.1 TL00272.10 TL00272.50
MAYBTL00272.1EA 0 EUR
MAYBTL00272.1 MAYBTL00272.10 MAYBTL00272.50
2,4,6-Trichlorobenzonitrile
2,4,6-Trichlorobenzonitrile
Formula: C₇H₂Cl₃N
MW: 206.46 g/mol
MDL Number: MFCD00052711
CAS Number: 6575-05-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR