4-Amino-2-chlorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Chloro-4-cyanoaniline
M-Clarity™ quality level = MQ200
279013-10G
279013-50G
SIAL279013-10GEA
86.2
EUR
SIAL279013-10G
SIAL279013-50G
4-Amino-2-chlorobenzonitrile, Sigma-Aldrich®
4-Amino-2-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Boiling Pt: 301 °C (1013 hPa) Melting Pt: 162…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035926 CAS Number: 20925-27-3 EINECS: 244-114-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...