5-Chloro-2-fluorobenzonitrile ≥97%, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
548693-1G 548693-5G
SIAL548693-1GEA 26.1 EUR
SIAL548693-1G SIAL548693-5G
5-Chloro-2-fluorobenzonitrile ≥97%, Sigma-Aldrich®
5-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 120 °C (20 -rr)
Melting Pt: 67…70 °C (lit.)
Density: 1,584 g/cm³ (20 °C)
MDL Number: MFCD01631434
CAS Number: 57381-34-7
EINECS: 252-132-5