2,4-Dichlorobenzoyl chloride
Supplier: Thermo Scientific
Danger
Synonyms: 2,4-Dichlorobenzene-1-carbonyl chloride
SB01809.1 SB01809.10 SB01809.50
MAYBSB01809.1EA 0 EUR
MAYBSB01809.1 MAYBSB01809.10 MAYBSB01809.50
2,4-Dichlorobenzoyl chloride
2,4-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00000661
CAS Number: 89-75-8
EINECS: 201-936-4
UN: 3265
ADR: 8,II