2-Chlorobenzoyl chloride 98%
Supplier: Thermo Scientific
Danger
Synonyms: o-Chlorobenzoyl chloride
147142500 147140010
ACRO147142500EA 70.6 EUR
ACRO147142500 ACRO147140010
2-Chlorobenzoyl chloride 98%
2-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: –4 °C
Density: 1.38 g/cm³
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00000660
CAS Number: 609-65-4
EINECS: 210-194-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5710 to 1.5750 (20°C, 589 nm)
GC ≥97.5 %
Titration RCOCl ≥97.5 %