2-Chlorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: o-Chlorobenzoyl chloride
M-Clarity™ quality level = MQ200
103918-100G 103918-500G 103918-5G
SIAL103918-100GEA 94 EUR
SIAL103918-100G SIAL103918-500G SIAL103918-5G
2-Chlorobenzoyl chloride, Sigma-Aldrich®
2-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000660
CAS Number: 609-65-4
EINECS: 210-194-0
UN: 3265
ADR: 8,II