3-Chlorobenzoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: m-Chlorobenzoyl chloride, 3-Chlorobenzoylchloride

M-Clarity™ quality level = MQ200

C26801-100G C26801-25G
SIALC26801-100GEA 90.3 EUR
SIALC26801-100G SIALC26801-25G
3-Chlorobenzoyl chloride, Sigma-Aldrich®
3-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 44 °C
Density: 1.367 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000671
CAS Number: 618-46-2
EINECS: 210-552-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR