3,5-Dichlorobenzamide

Supplier: Thermo Scientific

TL00896.1 TL00896.10 TL00896.50
MAYBTL00896.1EA 0 EUR
MAYBTL00896.1 MAYBTL00896.10 MAYBTL00896.50
3,5-Dichlorobenzamide
3,5-Dichlorobenzamide
Formula: C₇H₅Cl₂NO
MDL Number: MFCD00057644
CAS Number: 5980-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR