Methyl-2-chlorobenzoate
Supplier: Thermo Scientific
Synonyms: 2-Chlorobenzoic acid emthyl ester
TL00198.1 TL00198.10 TL00198.50
MAYBTL00198.1EA 0 EUR
MAYBTL00198.1 MAYBTL00198.10 MAYBTL00198.50
Methyl-2-chlorobenzoate
Methyl 2-chlorobenzoate
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Density: 1.247 g/cm³ (20 °C)
MDL Number: MFCD00016337
CAS Number: 610-96-8
EINECS: 210-242-0