Bottles, Narrow Neck, Round
Supplier: Rixius
RIXI1-0701-0050-22EA 219 EUR
RIXI1-0701-0050-22 RIXI1-0701-0010-18 RIXI1-0700-0015-20 RIXI1-0711-0015-22
Bottles, Narrow Neck, Round
Bottles Glass Bottles
Bottles, clear or amber glass, with thread.
  • Good chemical and physical properties
  • Stable, non-slip
  • For applications that do not involve high thermal stresses
Amber coloured to protect light-sensitive materials.
Informace pro objednávání: Screw caps to be ordered seperately.