2-Chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
135577-100G 135577-500G 135577-5G
SIAL135577-100GEA 44.6 EUR
SIAL135577-100G SIAL135577-500G SIAL135577-5G
2-Chlorobenzoic acid, Sigma-Aldrich®
2-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 285
Melting Pt: 139…142 °C
Density: 1.544 g/cm³ (20 °C)
Flash Pt: 173
Storage Temperature: Ambient
MDL Number: MFCD00002412
CAS Number: 118-91-2
EINECS: 204-285-4
Merck Index: 13,02143