3-Chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C24604-100G C24604-25G
SIALC24604-100GEA 60.9 EUR
SIALC24604-100G SIALC24604-25G
3-Chlorobenzoic acid, Sigma-Aldrich®
3-Chlorobenzoic acid
Formula: C₇H₅ClO₂
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 122 °C
Density: 1.496 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
EINECS: 208-618-4
Merck Index: 13,02142