2-Chloro-3-nitrobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning

141208-25G 141208-5G
SIAL141208-25GEA 238 EUR
SIAL141208-25G SIAL141208-5G
2-Chloro-3-nitrobenzoic acid, Sigma-Aldrich®
2-Chloro-3-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2
EINECS: 223-590-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR