2-Chloro-4-fluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

348074-1G 348074-5G
SIAL348074-1GEA 25 EUR
SIAL348074-1G SIAL348074-5G
2-Chloro-4-fluorobenzaldehyde, Sigma-Aldrich®
2-Chloro-4-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 60 °C
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR