2-Chloro-6-fluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
141240-100G 141240-25G
SIAL141240-100GEA 72.4 EUR
SIAL141240-100G SIAL141240-25G
2-Chloro-6-fluorobenzaldehyde, Sigma-Aldrich®
2-Chloro-6-fluorobenzaldehyde
Formula: C₇H₄ClFO
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003306
CAS Number: 387-45-1
EINECS: 206-860-5