3-Chloro-4-fluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
306002-1G
306002-5G
SIAL306002-1GEA
54.2
EUR
SIAL306002-1G
SIAL306002-5G
3-Chloro-4-fluorobenzaldehyde, Sigma-Aldrich®
3-Chloro-4-fluorobenzaldehyde
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 66 °C (2 mmHg) Melting Pt: 44…46 °C |
MDL Number:
MFCD00011735 CAS Number: 34328-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...