2-Chloro-4-fluorobenzamide

Supplier: Thermo Scientific

TL00229.1 TL00229.10
MAYBTL00229.1EA 0 EUR
MAYBTL00229.1 MAYBTL00229.10
2-Chloro-4-fluorobenzamide
2-Chloro-4-fluorobenzamide
Formula: C₇H₅ClFNO
MDL Number: MFCD00042531
CAS Number: 88578-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR