3-Chloro-4-fluorophenyl isothiocyanate
Supplier: Thermo Scientific
Synonyms:
1-Chloro-2-fluoro-5-isothiocyanatobenzene, 2-Chloro-1-fluoro-4-isothiocyanatobenzene
TL00696.1
TL00696.10
TL00696.50
MAYBTL00696.1EA
0
EUR
MAYBTL00696.1
MAYBTL00696.10
MAYBTL00696.50
3-Chloro-4-fluorophenyl isothiocyanate
3-Chloro-4-fluorophenyl isothiocyanate
Formula:
C₇H₃ClFNS |
MDL Number:
MFCD00060700 CAS Number: 137724-66-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...