2-Chloro-4-fluorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
162442-25G 162442-5G
SIAL162442-25GEA 102 EUR
SIAL162442-25G SIAL162442-5G
2-Chloro-4-fluorophenol, Sigma-Aldrich®
2-Chloro-4-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 23 °C
Density: 1.345 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002168
CAS Number: 1996-41-4
EINECS: 217-876-7
UN: 3265
ADR: 8,III