1-Chloro-3-iodobenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Chloroiodobenzene

M-Clarity™ quality level = MQ100

242578-5G
SIAL242578-5GEA 41.3 EUR
SIAL242578-5G
1-Chloro-3-iodobenzene, Sigma-Aldrich®
1-Chloro-3-iodobenzene
Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 54.5 °C
Density: 1.9255 g/cm³ (20 °C)
MDL Number: MFCD00001046
CAS Number: 625-99-0
EINECS: 210-920-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR