3,5-Dichlorophenyl isocyanate
Supplier: Thermo Scientific
Synonyms: 3,5-Dichlorophenylisocyantae
SB01467.1 SB01467.10 SB01467.50
MAYBSB01467.1EA 0 EUR
MAYBSB01467.1 MAYBSB01467.10 MAYBSB01467.50
3,5-Dichlorophenyl isocyanate
3,5-Dichlorophenyl isocyanate
Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 32…34 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013859
CAS Number: 34893-92-0
EINECS: 252-276-9
UN: 2250
ADR: 6.1,II