2,4-Dichloroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
112151-100G 112151-500G 112151-5G
SIAL112151-100GEA 53.3 EUR
SIAL112151-100G SIAL112151-500G SIAL112151-5G
2,4-Dichloroaniline, Sigma-Aldrich®
2,4-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 245
Melting Pt: 59…62 °C
Density: 1.567 g/cm³ (20 °C)
Flash Pt: 149
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1 II