2,6-Dichloroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D55504-100G D55504-25G D55504-500G
SIALD55504-100GEA 80.4 EUR
SIALD55504-100G SIALD55504-25G SIALD55504-500G
2,6-Dichloroaniline, Sigma-Aldrich®
2,6-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007675
CAS Number: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II